Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034129.1 | NP_001029301.1 | 7, 8 | 920 | ||
XM_006232224.2 | XP_006232286.1 | 3, 4 | 502 | |||
GenBank mRNA | AY325207.1 | - | 7, 8 | 920 | ||
FQ220436.1 | - | 3, 4 | 279 |