Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037183.1 | NP_001032260.1 | 7 | 1612 | ||
GenBank mRNA | BC089884.1 | - | 7 | 1613 | ||
BC105820.1 | - | 7 | 1612 | 7369885 |