Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TNNC2 SIRNA Assays ›
Gene Symbol:
Tnnc2
Gene Name:
troponin C type 2 (fast)
Gene Aliases:
-
Chromosome Location:
Chr.3: 161243747 - 161244831 on Build Rnor_6.0
UniGene ID:
Rn.43529
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
281554
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tnnc2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001037351.1
NP_001032428.1
3, 4
322
GenBank mRNA
DQ273679.1
-
3, 4
322
FQ215069.1
-
386
FQ216531.1
-
386
FQ216869.1
-
386
FQ216875.1
-
416
FQ216940.1
-
378
FQ216956.1
-
399
FQ217047.1
-
386
FQ217054.1
-
386
FQ217106.1
-
415
FQ217137.1
-
386
FQ217169.1
-
386
FQ217172.1
-
386
FQ217195.1
-
386
FQ217236.1
-
397
FQ217299.1
-
386
FQ217371.1
-
386
FQ217378.1
-
386
FQ217706.1
-
386
FQ217788.1
-
386
FQ217840.1
-
386
FQ217842.1
-
386
FQ217896.1
-
385
FQ217902.1
-
386
FQ217974.1
-
415
FQ217984.1
-
386
FQ218010.1
-
386
FQ218048.1
-
416
FQ218061.1
-
386
FQ223543.1
-
386
FQ223572.1
-
386
FQ223583.1
-
386
FQ223607.1
-
386
FQ223623.1
-
386
FQ223636.1
-
386
FQ223690.1
-
386
FQ223692.1
-
386
FQ223711.1
-
386
FQ223713.1
-
386
FQ223719.1
-
386
FQ223773.1
-
398
FQ223816.1
-
407
FQ223825.1
-
386
FQ223832.1
-
386
FQ223835.1
-
416
FQ223976.1
-
386
FQ223986.1
-
386
FQ224079.1
-
386
FQ224114.1
-
415
FQ224128.1
-
386
FQ224223.1
-
386
FQ224258.1
-
386
FQ224340.1
-
386
FQ224376.1
-
401
FQ224378.1
-
386
FQ224387.1
-
386
FQ224450.1
-
386
FQ224462.1
-
386
FQ224599.1
-
386
FQ224648.1
-
386
FQ224672.1
-
386
FQ224735.1
-
387
FQ224749.1
-
386
FQ224755.1
-
415
FQ224795.1
-
386
FQ224822.1
-
386
FQ224827.1
-
383
J00793.1
-
1, 2
106
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
skeletal muscle contraction
actin binding
calcium ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-97l6p:80/100.66.76.85:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline