Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ola1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033927.1 | NP_001029099.1 | 11 | 1430 | ||
GenBank mRNA | BC127457.1 | - | 11 | 1420 | 8363318 |