Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037767.2 | NP_001032856.2 | 4 | 605 | ||
GenBank mRNA | BC098820.1 | - | 3 | 746 | ||
BC127445.1 | - | 4 | 534 |