Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INPP5E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318502.1 | NP_001305431.1 | 10 | 2280 | ||
NM_019892.5 | NP_063945.2 | 10 | 2283 | |||
XM_017014926.1 | XP_016870415.1 | 10 | 2275 | |||
GenBank mRNA | AF187891.1 | - | 10 | 1984 | ||
AL832846.1 | - | 10 | 2281 | |||
BC028032.1 | - | 10 | 2265 | 5242186 | ||
U45974.1 | - | 4 | 497 |