Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASAH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143974.1 | NP_001137446.1 | 5 | 729 | ||
NM_019893.2 | NP_063946.2 | 5 | 729 | |||
XM_011539970.2 | XP_011538272.1 | 6 | 925 | |||
XM_011539971.2 | XP_011538273.1 | 6 | 929 | |||
XM_011539972.2 | XP_011538274.1 | 5 | 749 | |||
XM_011539973.2 | XP_011538275.1 | 6 | 935 | |||
XM_017016435.1 | XP_016871924.1 | 6 | 929 | |||
GenBank mRNA | AF250847.1 | - | 5 | 672 | ||
AF449759.2 | - | 5 | 729 | |||
BC107105.2 | - | 5 | 672 | 40018929 |