Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037544.1 | NP_001032633.1 | 11 | 1605 | ||
GenBank mRNA | AJ969954.1 | - | 11 | 1605 | ||
EF494736.1 | - | 1577 |