Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031648.1 | NP_001026818.1 | 4 | 1218 | ||
GenBank mRNA | BC098936.1 | - | 4 | 1218 | 7446346 |