Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ypel5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035221.1 | NP_001030298.1 | 598 | |||
XM_008764508.1 | XP_008762730.1 | 805 | ||||
XM_008764509.1 | XP_008762731.1 | 1053 | ||||
XM_008764510.1 | XP_008762732.1 | 1004 | ||||
GenBank mRNA | FQ225923.1 | - | 1 | 108 | ||
FQ229713.1 | - | 565 | ||||
FQ231463.1 | - | 1 | 108 | |||
FQ234574.1 | - | 1 | 108 |