Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034133.1 | NP_001029305.1 | 13 | 1295 | ||
GenBank mRNA | BC105756.1 | - | 13 | 1295 | 5623547 |