Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031649.1 | NP_001026819.1 | 8 | 1117 | ||
XM_006240896.1 | XP_006240958.1 | 9 | 1274 | |||
XM_006240897.2 | XP_006240959.1 | 8 | 1743 | |||
XM_008765091.1 | XP_008763313.1 | 8 | 1768 | |||
XM_008765092.1 | XP_008763314.1 | 9 | 1513 | |||
GenBank mRNA | BC098923.1 | - | 8 | 1117 | 7461534 |