Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020120.3 | NP_064505.1 | 12 | 1329 | ||
NR_027671.2 | 12 | 1482 | ||||
XM_006712634.3 | XP_006712697.1 | 12 | 1303 | |||
XM_006712635.3 | XP_006712698.1 | 12 | 1297 | |||
XM_006712636.3 | XP_006712699.1 | 12 | 1292 | |||
XM_017004507.1 | XP_016859996.1 | 13 | 1388 | |||
XM_017004508.1 | XP_016859997.1 | 13 | 1343 | |||
GenBank mRNA | AF227905.1 | - | 12 | 1189 | ||
AK293066.1 | - | 12 | 1482 | |||
AK309101.1 | - | 10 | 1054 | |||
BC041098.1 | - | 12 | 1479 | 5492334 |