Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txlng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037187.1 | NP_001032264.1 | 5 | 777 | ||
XM_006256890.2 | XP_006256952.1 | 5 | 837 | |||
XM_008773168.1 | XP_008771390.1 | 5 | 837 | |||
GenBank mRNA | BC099110.1 | - | 5 | 777 | 7461546 |