Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnjl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037773.4 | NP_001032862.2 | 5 | 1796 | ||
XM_006246133.2 | XP_006246195.1 | 6 | 2482 | |||
GenBank mRNA | BC099113.1 | - | 3 | 1428 |