Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBQLN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304342.1 | NP_001291271.1 | 11 | 2615 | ||
NM_020131.4 | NP_064516.2 | 11 | 2675 | |||
GenBank mRNA | AB209566.1 | - | 11 | 2548 | ||
AF113544.1 | - | 8 | 2081 | |||
AF188240.1 | - | 11 | 2829 | |||
AK098368.1 | - | 6 | 1520 | |||
BC006410.1 | - | 2 | 950 | |||
BC018403.2 | - | 11 | 2563 | 4214386 | ||
BC063841.1 | - | 11 | 2575 | 6183942 |