Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037645.1 | NP_001032734.1 | 8 | 869 | ||
XM_006251883.2 | XP_006251945.1 | 9 | 969 | |||
GenBank mRNA | BC092636.1 | - | 3 | 384 | ||
BC107443.1 | - | 8 | 869 | 7307807 |