Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csf1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029901.1 | NP_001025072.1 | 22 | 2744 | ||
XM_006254813.2 | XP_006254875.1 | 19 | 2842 | |||
XM_008772147.1 | XP_008770369.1 | 20 | 2869 | |||
XM_008772148.1 | XP_008770370.1 | 20 | 2891 | |||
GenBank mRNA | X61479.1 | - | 19 | 2687 |