Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc45a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037647.1 | NP_001032736.1 | 8 | 1032 | ||
XM_006229432.2 | XP_006229494.1 | 8 | 983 | |||
GenBank mRNA | BC107919.1 | - | 8 | 1032 | 7457637 |