Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dctn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037778.1 | NP_001032867.1 | 6 | 1562 | ||
GenBank mRNA | BC098764.1 | - | 6 | 1562 | 7371224 |