Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035251.1 | NP_001030328.1 | 3 | 371 | ||
XM_006231929.2 | XP_006231991.1 | 4 | 498 | |||
XM_006231930.2 | XP_006231992.1 | 5 | 562 | |||
XM_006231931.2 | XP_006231993.1 | 4 | 539 | |||
XM_006231932.2 | XP_006231994.1 | 4 | 629 | |||
XM_006231933.2 | XP_006231995.1 | 4 | 588 | |||
XM_006231934.2 | XP_006231996.1 | 3 | 2573 | |||
XM_006231936.2 | XP_006231998.1 | 3 | 2558 | |||
XM_006231937.2 | XP_006231999.1 | 3 | 339 | |||
XM_006231939.2 | XP_006232001.1 | 3 | 251 | |||
GenBank mRNA | BC092662.1 | - | 2 | 233 | ||
BC105869.1 | - | 3 | 371 | 7442690 | ||
JQ013729.1 | - | 3 | 422 |