Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K7CL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286617.1 | NP_001273546.1 | 10 | 2093 | ||
NM_001286618.1 | NP_001273547.1 | 9 | 1964 | |||
NM_001286619.1 | NP_001273548.1 | 7 | 1904 | |||
NM_001286620.1 | NP_001273549.1 | 5 | 1742 | |||
NM_001286622.1 | NP_001273551.1 | 9 | 1999 | |||
NM_001286624.1 | NP_001273553.1 | 4 | 1343 | |||
NM_001286634.1 | NP_001273563.1 | 8 | 1855 | |||
NM_020152.3 | NP_064537.1 | 10 | 2084 | |||
GenBank mRNA | AF269161.1 | - | 8 | 1693 | ||
AF269162.1 | - | 10 | 1931 | |||
AF269163.1 | - | 9 | 1802 | |||
AF269164.1 | - | 5 | 1531 | |||
AK056341.1 | - | 5 | 1583 | |||
AK122780.1 | - | 7 | 1904 | |||
AL713701.1 | - | 5 | 1552 | |||
AY171599.2 | - | 9 | 1841 | |||
EF219158.1 | - | 4 | 1354 | |||
EF219159.1 | - | 5 | 1393 | |||
EF219160.1 | - | 4 | 1304 |