Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FHIT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166243.2 | NP_001159715.1 | 8 | 686 | ||
NM_001320899.1 | NP_001307828.1 | 8 | 686 | |||
NM_001320900.1 | NP_001307829.1 | 9 | 780 | |||
NM_001320901.1 | NP_001307830.1 | 6 | 610 | |||
NM_002012.3 | NP_002003.1 | 8 | 686 | |||
NR_135491.1 | 5 | 627 | ||||
XM_017005880.1 | XP_016861369.1 | 9 | 734 | |||
XM_017005881.1 | XP_016861370.1 | 8 | 696 | |||
XM_017005882.1 | XP_016861371.1 | 9 | 719 | |||
XM_017005883.1 | XP_016861372.1 | 9 | 734 | |||
XM_017005884.1 | XP_016861373.1 | 9 | 734 | |||
XM_017005885.1 | XP_016861374.1 | 5 | 641 | |||
XM_017005886.1 | XP_016861375.1 | 5 | 1401 | |||
GenBank mRNA | AK127931.1 | - | 5 | 340 | ||
AK289824.1 | - | 8 | 684 | |||
BC032336.1 | - | 8 | 638 | 5185162 | ||
DB131595.1 | - | 6 | 322 | |||
DQ120721.1 | - | 4 | 313 | |||
KJ534835.1 | - | 4 | 313 | |||
U46922.1 | - | 8 | 675 |