Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rexo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033884.1 | NP_001029056.1 | 8 | 2092 | ||
GenBank mRNA | BC161820.1 | - | 8 | 2057 | 9088929 |