Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ing3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034107.1 | NP_001029279.1 | 11, 12 | 1219 | ||
XM_006236140.2 | XP_006236202.1 | 11, 12 | 1261 | |||
XM_006236141.2 | XP_006236203.1 | 10, 11 | 1222 | |||
GenBank mRNA | BC100082.1 | - | 11, 12 | 1219 | 7385392 | |
FQ232896.1 | - | 2, 3 | 135 |