Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Immt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034928.1 | NP_001030100.1 | 9 | 1007 | ||
XM_008762981.1 | XP_008761203.1 | 10 | 1237 | |||
XM_008762982.1 | XP_008761204.1 | 10 | 1237 | |||
XM_008762983.1 | XP_008761205.1 | 10 | 1203 | |||
XM_008762984.1 | XP_008761206.1 | 10 | 1200 | |||
XM_008762985.1 | XP_008761207.1 | 10 | 1203 | |||
XM_008762986.1 | XP_008761208.1 | 9 | 1139 | |||
XM_008762987.1 | XP_008761209.1 | 9 | 1105 | |||
XM_008762988.1 | XP_008761210.1 | 9 | 1102 | |||
XM_008762989.1 | XP_008761211.1 | 9 | 1105 | |||
XM_008762990.1 | XP_008761212.1 | 8 | 1012 | |||
GenBank mRNA | BC083835.1 | - | 962 | |||
BC105841.1 | - | 9 | 1007 | 7454026 | ||
FQ217458.1 | - | 8 | 934 |