Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serinc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031656.1 | NP_001026826.1 | 7, 8 | 917 | ||
GenBank mRNA | BC099093.1 | - | 7, 8 | 917 | 7457983 | |
DQ103709.1 | - | 7, 8 | 903 |