Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paqr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034081.1 | NP_001029253.1 | 1 | 170 | ||
XM_006239130.2 | XP_006239192.1 | 2 | 268 | |||
XM_008764145.1 | XP_008762367.1 | 3 | 491 | |||
XM_008764146.1 | XP_008762368.1 | 3 | 279 | |||
GenBank mRNA | DQ027002.1 | - | 1 | 170 |
Set Membership: |
G-protein coupled receptor |