Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037354.1 | NP_001032431.1 | 4 | 2060 | ||
GenBank mRNA | DQ232764.2 | - | 4 | 2060 |