Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034150.1 | NP_001029322.1 | 3 | 348 | ||
GenBank mRNA | BC105782.1 | - | 3 | 348 | 5599400 |