Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xylb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033704.1 | NP_001028876.1 | 18 | 1473 | ||
XM_006244093.1 | XP_006244155.1 | 19 | 1845 | |||
XM_006244096.1 | XP_006244158.1 | 17 | 1485 | |||
GenBank mRNA | BC101852.1 | - | 18 | 1473 | 7930657 |