Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xylb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033704.1 | NP_001028876.1 | 10 | 786 | ||
XM_006244093.1 | XP_006244155.1 | 11 | 1158 | |||
XM_006244094.1 | XP_006244156.1 | 11 | 1158 | |||
XM_006244095.1 | XP_006244157.1 | 11 | 1158 | |||
XM_006244096.1 | XP_006244158.1 | 9 | 798 | |||
GenBank mRNA | BC101852.1 | - | 10 | 786 | 7930657 |