Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321463.1 | NP_001308392.1 | 15 | 1836 | ||
NM_020170.3 | NP_064555.2 | 15 | 1839 | |||
GenBank mRNA | AK125002.1 | - | 15 | 1895 | ||
AK172848.1 | - | 15 | 1786 | |||
AL136567.1 | - | 15 | 1670 | |||
AL365369.1 | - | 3 | 182 | |||
BC003076.2 | - | 7 | 526 | |||
BC007275.1 | - | 9 | 815 | |||
BC008920.2 | - | 11 | 1046 | |||
BC010064.2 | - | 15 | 1755 | 4100399 | ||
BC013283.2 | - | 15 | 2116 | 3959506 | ||
BC019091.2 | - | 15 | 1755 | 4895124 | ||
BC025926.1 | - | 15 | 1748 | 3509966 | ||
CR533566.1 | - | 15 | 1652 |