Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc11a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031658.1 | NP_001026828.1 | 14 | 1781 | ||
XM_006245219.2 | XP_006245281.1 | 15 | 1859 | |||
XM_006245220.1 | XP_006245282.1 | 15 | 1907 | |||
GenBank mRNA | BC100653.1 | - | 14 | 1781 | 7932285 |