Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sft2d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034011.1 | NP_001029183.1 | 8 | 1995 | ||
GenBank mRNA | BC099092.1 | - | 8 | 1995 | 7457938 |