Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tada1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037980.2 | NP_001033069.2 | 8 | 1055 | ||
GenBank mRNA | BC091380.1 | - | 847 | 7320760 | ||
BC158713.1 | - | 8 | 1055 | 9035442 |