Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNNM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020184.3 | NP_064569.3 | 7 | 2746 | ||
XM_005263914.3 | XP_005263971.1 | 8 | 2726 | |||
XM_005263915.3 | XP_005263972.1 | 7 | 2751 | |||
XM_017003799.1 | XP_016859288.1 | 7 | 1322 | |||
GenBank mRNA | AB046812.1 | - | 7 | 2476 | ||
AF202777.1 | - | 7 | 2507 | |||
AK022833.1 | - | 4 | 941 | |||
AK095065.1 | - | 1 | 809 | |||
BC063295.1 | - | 7 | 2746 | 30340626 |