Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033068.3 | NP_001028240.3 | 17 | 2256 | ||
GenBank mRNA | BC103628.1 | - | 17 | 2256 | 7939290 |