Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193557.1 | NP_001180486.1 | 5 | 337 | ||
NM_020189.5 | NP_064574.1 | 5 | 409 | |||
NR_036471.1 | 5 | 407 | ||||
NR_036472.1 | 5 | 454 | ||||
GenBank mRNA | AF173296.1 | - | 5 | 479 | ||
AF201940.1 | - | 5 | 424 | |||
AK000487.1 | - | 5 | 313 | |||
AK095651.1 | - | 5 | 453 | |||
AK290752.1 | - | 5 | 353 | |||
AK316560.1 | - | 5 | 404 | |||
BC007870.2 | - | 5 | 325 | 4303290 | ||
BX537648.1 | - | 2 | 233 | |||
BX648621.1 | - | 3 | 1158 | |||
CR457183.1 | - | 4 | 263 | |||
DA548665.1 | - | 5 | 283 | |||
DA762166.1 | - | 5 | 389 | |||
DB168034.1 | - | 5 | 409 |