Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YAE1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282446.1 | NP_001269375.1 | 2 | 256 | ||
NM_020192.4 | NP_064577.1 | 2 | 256 | |||
GenBank mRNA | AF226046.1 | - | 2 | 222 | ||
AK293511.1 | - | 2 | 226 | |||
AK314937.1 | - | 2 | 228 | |||
BC022043.1 | - | 2 | 226 | 4734433 | ||
BM767075.1 | - | 2 | 256 | |||
BX648555.1 | - | 1 | 1162 | |||
CR457198.1 | - | 2 | 213 |