Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EMSY SIRNA Assays ›
Gene Symbol:
EMSY
Gene Name:
EMSY, BRCA2 interacting transcriptional repressor
Gene Aliases:
C11orf30, GL002
Chromosome Location:
Chr.11: 76444928 - 76552901 on Build GRCh38
UniGene ID:
Hs.352588
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
28215
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EMSY
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001300942.1
NP_001287871.1
14, 15
2191
NM_001300943.1
NP_001287872.1
13, 14
2149
NM_001300944.1
NP_001287873.1
14, 15
2191
NM_020193.4
NP_064578.2
13, 14
2146
XM_005274112.4
XP_005274169.1
13, 14
2135
XM_006718637.3
XP_006718700.1
14, 15
2288
XM_006718638.3
XP_006718701.1
13, 14
2159
XM_006718639.3
XP_006718702.1
12, 13
2006
XM_011545173.2
XP_011543475.1
14, 15
2288
XM_011545174.2
XP_011543476.1
14, 15
2285
XM_011545175.2
XP_011543477.1
13, 14
2159
XM_011545177.2
XP_011543479.1
13, 14
2135
XM_011545178.2
XP_011543480.1
14, 15
2288
XM_011545179.2
XP_011543481.1
12, 13
2006
XM_017018021.1
XP_016873510.1
14, 15
2288
XM_017018022.1
XP_016873511.1
14, 15
2134
GenBank mRNA
AJ430203.1
-
12, 13
2003
AK125114.1
-
12, 13
1860
AK126030.1
-
1, 2
165
AK304043.1
-
10, 11
1555
AK304106.1
-
1, 2
56
BC117265.1
-
12, 13
2045
40125816
BC143370.1
-
13, 14
2084
9051875
BC143374.1
-
13, 14
2087
9051879
BC143376.1
-
13, 14
2087
9051881
BX647398.1
-
2, 3
241
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
DNA repair
transcription, DNA-templated
regulation of transcription, DNA-templated
covalent chromatin modification
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-xzn5f:80/10.239.160.173:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline