Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Carm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030041.3 | NP_001025212.1 | 6 | 896 | ||
NM_001034088.2 | NP_001029260.1 | 6 | 896 | |||
GenBank mRNA | AB201114.1 | - | 6 | 896 | ||
AB201115.1 | - | 6 | 896 | |||
AB201116.1 | - | 6 | 896 | |||
AB201117.1 | - | 6 | 896 | |||
KJ672502.1 | - | 6 | 806 |