Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195286.1 | NP_001182215.1 | 2, 3 | 350 | ||
NM_002460.3 | NP_002451.2 | 2, 3 | 350 | |||
NR_046000.2 | 2, 3 | 350 | ||||
XM_006715090.2 | XP_006715153.1 | 2, 3 | 337 | |||
GenBank mRNA | AK310149.1 | - | 2, 3 | 337 | ||
BC015752.1 | - | 2, 3 | 297 | 4861223 | ||
D78261.1 | - | 2, 3 | 319 | |||
U52682.1 | - | 2, 3 | 349 | |||
U63738.1 | - | 2, 3 | 329 |