Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1311300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF196199.1 | - | 6 | 884 | ||
AF196210.1 | - | 6 | 782 | |||
AF196212.1 | - | 6 | 782 | |||
AF196213.1 | - | 5 | 774 | |||
AF196215.1 | - | 5 | 785 | |||
AF196216.1 | - | 5 | 785 | |||
AF196217.1 | - | 821 | ||||
AF196218.1 | - | 828 | ||||
AF196219.1 | - | 821 | ||||
AF196220.1 | - | 821 | ||||
AF196221.1 | - | 825 | ||||
AF196222.1 | - | 824 | ||||
AF196223.1 | - | 740 | ||||
AF196224.1 | - | 839 | ||||
AF196225.1 | - | 853 | ||||
AF196226.1 | - | 861 | ||||
AF196227.1 | - | 715 | ||||
AF196228.1 | - | 825 | ||||
AF196229.1 | - | 7 | 757 | |||
AF196230.1 | - | 5 | 542 | |||
AF196231.1 | - | 6 | 853 | |||
AF196232.1 | - | 8 | 756 | |||
AF196233.1 | - | 6 | 774 | |||
AF196234.1 | - | 6 | 838 | |||
AF196235.1 | - | 6 | 664 | |||
AF196236.1 | - | 6 | 864 | |||
AF196237.1 | - | 7 | 743 | |||
AF196238.1 | - | 8 | 767 | |||
AF196239.1 | - | 7 | 885 | |||
AF196240.1 | - | 7 | 771 | |||
AF196242.1 | - | 6 | 849 | |||
AF196243.1 | - | 7 | 745 | |||
AF196244.1 | - | 8 | 734 | |||
AF196245.1 | - | 7 | 823 | |||
AF196246.1 | - | 5 | 793 | |||
AF196247.1 | - | 8 | 734 | |||
AF196266.1 | - | 4 | 443 | |||
AF196267.1 | - | 2 | 66 | |||
AJ249228.1 | - | 6 | 529 | |||
BC099216.1 | - | 6 | 884 | 7384680 |