Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034943.1 | NP_001030115.1 | 7 | 1382 | ||
XM_008760161.1 | XP_008758383.1 | 8 | 2549 | |||
XM_008760162.1 | XP_008758384.1 | 8 | 2397 | |||
GenBank mRNA | BC103638.1 | - | 7 | 1382 | 7109619 |