Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020201.3 | NP_064586.1 | 2 | 466 | ||
XM_005256731.3 | XP_005256788.1 | 2 | 455 | |||
XM_005256732.3 | XP_005256789.1 | 2 | 455 | |||
XM_005256733.3 | XP_005256790.1 | 2 | 455 | |||
XM_005256734.3 | XP_005256791.1 | 2 | 455 | |||
XM_005256737.1 | XP_005256794.1 | 3 | 268 | |||
XM_011523962.2 | XP_011522264.1 | 2 | 455 | |||
XM_011523963.1 | XP_011522265.1 | 3 | 212 | |||
XM_011523964.2 | XP_011522266.1 | 3 | 181 | |||
XM_011523965.2 | XP_011522267.1 | 2 | 85 | |||
XM_011523966.1 | XP_011522268.1 | 2 | 455 | |||
XM_011523967.2 | XP_011522269.1 | 2 | 455 | |||
XM_017024879.1 | XP_016880368.1 | 2 | 1115 | |||
GenBank mRNA | AF210652.1 | - | 2 | 497 | ||
AL526346.3 | - | 2 | 550 | |||
BC035838.1 | - | 2 | 440 | 5727827 | ||
DQ323995.1 | - | 2 | 281 |