Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clul1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033071.1 | NP_001028243.1 | 8, 9 | 1256 | ||
XM_008767426.1 | XP_008765648.1 | 4, 5 | 1048 | |||
XM_008767427.1 | XP_008765649.1 | 4, 5 | 1003 | |||
GenBank mRNA | AY655707.1 | - | 8, 9 | 1256 |