Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034945.1 | NP_001030117.1 | 4 | 824 | ||
XM_006249464.2 | XP_006249526.1 | 4 | 841 | |||
GenBank mRNA | BC103639.1 | - | 4 | 824 | 7133734 |