Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idnk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037362.1 | NP_001032439.1 | 5 | 744 | ||
XM_006253563.2 | XP_006253625.1 | 4 | 875 | |||
XM_006253564.2 | XP_006253626.1 | 4 | 749 | |||
GenBank mRNA | BC087713.1 | - | 4 | 711 | ||
BC107923.1 | - | 5 | 744 | 7444531 |