Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhga12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037337.1 | NP_001032414.1 | 1 | 1581 | ||
GenBank mRNA | AY574011.1 | - | 1 | 1581 |